ENA372574070 Enamine

methyl 1-cyclopentylpyrrolidine-2-carboxylate

  • MDL number MFCD16160202
  • Cas Number : 1485728-95-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114370

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114370
Availability details for
Quantity
  • Availability
  • Add to Cart