ENA372574075 Enamine

O-(3,3,3-trifluoropropyl)hydroxylamine hydrochloride

  • Molecular Weight : 165.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(aminooxy)-1,1,1-trifluoropropane hydrochloride
Inchi Code 1S/C3H6F3NO.ClH/c4-3(5,6)1-2-8-7;/h1-2,7H2;1H
Inchi Key GBVOLUQNPMGCOH-UHFFFAOYSA-N

Description

Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-114376
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart