ENA372574077 Enamine

3-chloro-6-[3-(trifluoromethyl)phenyl]pyridazine

  • MDL number MFCD12439035
  • Cas Number : 66548-63-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114380

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114380
Availability details for
Quantity
  • Availability
  • Add to Cart