ENA372574083 Enamine

N-[(1-benzofuran-2-yl)methyl]cyclopentanamine

  • MDL number MFCD11157439
  • Cas Number : 1038232-05-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114419

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114419
Availability details for
Quantity
  • Availability
  • Add to Cart