ENA372574086 Enamine

6-methyl-1H,6H,7H-pyrazolo[3,4-d]pyridazin-7-one

  • MDL number MFCD16619851
  • Cas Number : 69792-72-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114429

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114429
Availability details for
Quantity
  • Availability
  • Add to Cart