ENA372574087 Enamine

4-chloro-N,N-dimethyl-3-sulfamoylbenzamide

  • MDL number MFCD11650002
  • Cas Number : 90561-59-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130...132
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114435

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114435
Availability details for
Quantity
  • Availability
  • Add to Cart