ENA372574089 Enamine

2-(6-chloro-2-methylpyridin-3-yl)acetic acid

  • MDL number MFCD09925308
  • Cas Number : 1000568-04-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134...136
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114442

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114442
Availability details for
Quantity
  • Availability
  • Add to Cart