ENA372574099 Enamine

1,3-difluoro-2,4-dinitrobenzene

  • MDL number MFCD00798573
  • Cas Number : 2106-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113215

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113215
Availability details for
Quantity
  • Availability
  • Add to Cart