ENA372574101 Enamine

2-[(2-oxooxolan-3-yl)sulfanyl]acetic acid

  • MDL number MFCD11646506
  • Cas Number : 1094479-23-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113218

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113218
Availability details for
Quantity
  • Availability
  • Add to Cart