ENA372574114 Enamine

2-chloro-1-(2-methyl-1,2,3,4-tetrahydroquinolin-1-yl)propan-1-one

  • MDL number MFCD09930466
  • Cas Number : 78142-21-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28175

Safety Information

msds https://www.enaminestore.com/catalog/EN300-28175
Availability details for
Quantity
  • Availability
  • Add to Cart