ENA372574115 Enamine

1-(5-methoxy-2-methyl-2,3-dihydro-1-benzofuran-6-yl)methanamine

  • MDL number MFCD01245878
  • Cas Number : 314028-47-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28286

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28286
Availability details for
Quantity
  • Availability
  • Add to Cart