ENA372574117 Enamine

2-bromo-N-(butan-2-yl)-3-methylbutanamide

  • MDL number MFCD01070260
  • Cas Number : 1016740-90-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29017

Safety Information

msds https://www.enaminestore.com/catalog/EN300-29017
Availability details for
Quantity
  • Availability
  • Add to Cart