ENA372574122 Enamine

2-bromo-N-(2-methoxyphenyl)-3-methylbutanamide

  • MDL number MFCD04406114
  • Cas Number : 391220-13-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29041

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29041
Availability details for
Quantity
  • Availability
  • Add to Cart