ENA372574127 Enamine

2-bromo-N-cycloheptyl-3-methylbutanamide

  • MDL number MFCD11643987
  • Cas Number : 1098361-68-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29083

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29083
Availability details for
Quantity
  • Availability
  • Add to Cart