ENA372574130 Enamine

2-bromo-N-[(2-fluorophenyl)methyl]-3-methylbutanamide

  • MDL number MFCD09932641
  • Cas Number : 1016726-55-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104...106
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29116

Safety Information

msds https://www.enaminestore.com/catalog/EN300-29116
Availability details for
Quantity
  • Availability
  • Add to Cart