ENA372574133 Enamine

2-bromo-3-methyl-1-(1,2,3,4-tetrahydroquinolin-1-yl)butan-1-one

  • MDL number MFCD09942473
  • Cas Number : 92378-02-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29140

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29140
Availability details for
Quantity
  • Availability
  • Add to Cart