ENA372574134 Enamine

2-bromo-3-methyl-1-(pyrrolidin-1-yl)butan-1-one

  • MDL number MFCD09951043
  • Cas Number : 1017021-12-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29143

Safety Information

msds https://www.enaminestore.com/catalog/EN300-29143
Availability details for
Quantity
  • Availability
  • Add to Cart