ENA372574136 Enamine

2-bromo-1-(3,5-dimethylpiperidin-1-yl)-3-methylbutan-1-one

  • MDL number MFCD09946081
  • Cas Number : 1134516-21-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29148

Safety Information

msds https://www.enaminestore.com/catalog/EN300-29148
Availability details for
Quantity
  • Availability
  • Add to Cart