ENA372574138 Enamine

2-bromo-1-(3,4-dihydro-2H-1,4-benzothiazin-4-yl)-3-methylbutan-1-one

  • MDL number MFCD11643992
  • Cas Number : 1097794-82-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29160

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29160
Availability details for
Quantity
  • Availability
  • Add to Cart