ENA372574145 Enamine

3-hydrazinyl-2-methylpropanenitrile

  • MDL number MFCD22053192
  • Cas Number : 352-16-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36530

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36530
Availability details for
Quantity
  • Availability
  • Add to Cart