ENA372574148 Enamine

N-[(3-nitrophenyl)methyl]cyclopropanamine

  • MDL number MFCD07408042
  • Cas Number : 697305-95-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41931

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41931
Availability details for
Quantity
  • Availability
  • Add to Cart