ENA372574149 Enamine

2,4-dichloro-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD09909676
  • Cas Number : 62019-56-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164...166
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42298

Safety Information

msds https://www.enaminestore.com/catalog/EN300-42298
Availability details for
Quantity
  • Availability
  • Add to Cart