ENA372574152 Enamine

1-tert-butyl-4,5,6,7-tetrahydro-1H-indazol-4-amine

  • MDL number MFCD21103118
  • Cas Number : 1181826-14-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44098

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44098
Availability details for
Quantity
  • Availability
  • Add to Cart