ENA372574155 Enamine

3-(2,3-dihydro-1-benzofuran-5-yl)-1,2-oxazole-5-carboxylic acid

  • MDL number MFCD12065173
  • Cas Number : 1157546-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 235...237
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113222

Safety Information

msds https://www.enaminestore.com/catalog/EN300-113222
Availability details for
Quantity
  • Availability
  • Add to Cart