ENA372574162 Enamine

5-(2,2-difluoroethoxy)pyridine-3-carboxylic acid

  • MDL number MFCD18898219
  • Cas Number : 1342512-20-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-112914

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112914
Availability details for
Quantity
  • Availability
  • Add to Cart