ENA372574168 Enamine

2-(chloromethyl)-5-(ethanesulfonyl)-1,3-benzoxazole

  • MDL number MFCD09880542
  • Cas Number : 1018186-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-50005

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50005
Availability details for
Quantity
  • Availability
  • Add to Cart