ENA372574178 Enamine

2,2,2-trifluoroethyl N-(1,3-dioxo-2,3-dihydro-1H-isoindol-5-yl)carbamate

  • MDL number MFCD22392242
  • Cas Number : 1394777-21-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-56658

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56658
Availability details for
Quantity
  • Availability
  • Add to Cart