ENA372574188 Enamine

4-nitro-5-(propan-2-yl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD09052668
  • Cas Number : 141721-97-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149...151
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60601

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60601
Availability details for
Quantity
  • Availability
  • Add to Cart