ENA372574190 Enamine

oxane-2-carboxylic acid

  • MDL number MFCD07779239
  • Cas Number : 51673-83-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61323

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-61323
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart