ENA372574191 Enamine

4-chloro-2-methyl-6-(pyridin-3-yl)pyrimidine

  • MDL number MFCD12032904
  • Cas Number : 1159817-95-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-63620

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63620
Availability details for
Quantity
  • Availability
  • Add to Cart