ENA372574206 Enamine

3-methoxy-5H,6H,7H,8H-pyrido[4,3-c]pyridazine

  • MDL number MFCD22392245
  • Cas Number : 1394645-71-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70875

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70875
Availability details for
Quantity
  • Availability
  • Add to Cart