ENA372574213 Enamine

2-(3-methylpyrrolidin-1-yl)ethan-1-ol

  • MDL number MFCD14601221
  • Cas Number : 126512-75-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113239

Safety Information

msds https://www.enaminestore.com/catalog/EN300-113239
Availability details for
Quantity
  • Availability
  • Add to Cart