ENA372574227 Enamine

5-amino-1H-pyrazole-4-carbothioamide

  • MDL number MFCD07364430
  • Cas Number : 34954-13-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80541

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80541
Availability details for
Quantity
  • Availability
  • Add to Cart