ENA372574228 Enamine

ethyl 2,2-dimethylpropanecarboximidate hydrochloride

  • MDL number MFCD00196042
  • Cas Number : 58910-26-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116...118
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80780

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80780
Availability details for
Quantity
  • Availability
  • Add to Cart