ENA372574264 Enamine

1-[5-(trifluoromethyl)pyridin-2-yl]-1H-pyrazol-5-amine

  • MDL number MFCD11641440
  • Cas Number : 107748-59-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112925

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112925
Availability details for
Quantity
  • Availability
  • Add to Cart