ENA372574266 Enamine

5-bromo-2-methylthiophene-3-sulfonyl chloride

  • MDL number MFCD21231989
  • Cas Number : 82834-48-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91696

Safety Information

signalword Danger
pcodes P260,P261,P264,P270,P271,P280,P301+P312,P301+P330+P331,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P330,P363,P405,P501
hcodes C,Xn
MSDS https://www.enaminestore.com/catalog/EN300-91696
pictograms GHS05,GHS07
hstatements H302,H312,H314,H332
Availability details for
Quantity
  • Availability
  • Add to Cart