ENA372574272 Enamine

3-methanesulfonyl-2-methylaniline

  • MDL number MFCD17284865
  • Cas Number : 1335496-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117...119
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112928

Safety Information

msds https://www.enaminestore.com/catalog/EN300-112928
Availability details for
Quantity
  • Availability
  • Add to Cart