ENA372574293 Enamine

(4-methyl-5-sulfanyl-4H-1,2,4-triazol-3-yl)methanol

  • MDL number MFCD18818084
  • Cas Number : 65029-36-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189...191
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97317

Safety Information

msds https://www.enaminestore.com/catalog/EN300-97317
Availability details for
Quantity
  • Availability
  • Add to Cart