ENA375227307 Enamine

methyl 2-hydroxy-2-(piperidin-4-yl)acetate hydrochloride

  • MDL number MFCD22565847
  • Cas Number : 1423026-48-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 141...143
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107730

Safety Information

msds https://www.enaminestore.com/catalog/EN300-107730
Availability details for
Quantity
  • Availability
  • Add to Cart