ENA375227312 Enamine

5-tert-butylthiophene-2-sulfonamide

  • MDL number MFCD22565851
  • Cas Number : 142294-56-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-108959

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108959
Availability details for
Quantity
  • Availability
  • Add to Cart