ENA375227314 Enamine

4-carbamoyl-3-ethyl-5-methyl-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD22565854
  • Cas Number : 1423029-63-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109527

Safety Information

msds https://www.enaminestore.com/catalog/EN300-109527
Availability details for
Quantity
  • Availability
  • Add to Cart