ENA375227322 Enamine

1-cyclopropyl-1H-imidazo[4,5-c]pyridine

  • MDL number MFCD22565864
  • Cas Number : 1423026-30-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53...55
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112950

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-112950
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart