ENA375227323 Enamine

2-(3-oxocycloheptyl)-2,3-dihydro-1H-isoindole-1,3-dione

  • MDL number MFCD22565865
  • Cas Number : 1423029-25-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113-115
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113198

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113198
Availability details for
Quantity
  • Availability
  • Add to Cart