ENA375227324 Enamine

4-chloro-2-(1-methyl-1H-imidazol-2-yl)pyridine

  • MDL number MFCD22565867
  • Cas Number : 1423030-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113337

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113337
Availability details for
Quantity
  • Availability
  • Add to Cart