ENA375227329 Enamine

5-(methoxymethyl)-3-methyl-1H-1,2,4-triazole hydrochloride

  • MDL number MFCD22565872
  • Cas Number : 1423024-01-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135...137
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114030

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114030
Availability details for
Quantity
  • Availability
  • Add to Cart