ENA375227334 Enamine

2,3-dimethyl 7-azabicyclo[2.2.1]heptane-2,3-dicarboxylate

  • MDL number MFCD22565877
  • Cas Number : 1393477-05-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77...79
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114220

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114220
Availability details for
Quantity
  • Availability
  • Add to Cart