ENA375227335 Enamine

ethyl 2,2-difluoro-2-(3-oxocyclopentyl)acetate

  • MDL number MFCD22565878
  • Cas Number : 1423034-15-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114221

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114221
Availability details for
Quantity
  • Availability
  • Add to Cart