ENA375227336 Enamine

tert-butyl N-(2-methyl-1,3-thiazol-5-yl)carbamate

  • MDL number MFCD22565879
  • Cas Number : 936361-25-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147...149
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114228

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114228
Availability details for
Quantity
  • Availability
  • Add to Cart