ENA375227338 Enamine

2-[(cyclopropylmethyl)amino]-2-(3,5-dichlorophenyl)acetic acid hydrochloride

  • MDL number MFCD22565881
  • Cas Number : 1423025-21-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 239...241
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114276

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114276
Availability details for
Quantity
  • Availability
  • Add to Cart