ENA375227345 Enamine

5-(aminomethyl)-2-methoxybenzonitrile hydrochloride

  • MDL number MFCD22565888
  • Cas Number : 1423032-10-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 249...251
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114449

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114449
Availability details for
Quantity
  • Availability
  • Add to Cart